Thermo Scientific Chemicals

Methyl 3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 168440250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Methyl 3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 168440250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 168440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
618-95-1
IUPAC Name
methyl 3-nitrobenzoate
Molecular Formula
C8H7NO4
InChI Key
AXLYJLKKPUICKV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
76°C to 80°C
Appearance (Color)
Beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text