1,1'-Thiocarbonyldiimidazole, 90%, Tech., Thermo Scientific Chemicals
1,1'-Thiocarbonyldiimidazole, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Thiocarbonyldiimidazole, 90%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number 168480250
Price (USD)/ Each
175.65
Online exclusive
206.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
175.65
Online exclusive
206.00 
Save 30.35 (15%)
Add to cart
1,1'-Thiocarbonyldiimidazole, 90%, Tech., Thermo Scientific Chemicals
Catalog number168480250
Price (USD)/ Each
175.65
Online exclusive
206.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-(1H-imidazole-1-carbothioyl)-1H-imidazole
Molecular FormulaC7H6N4S
InChI KeyRAFNCPHFRHZCPS-UHFFFAOYSA-N
SMILESS=C(N1C=CN=C1)N1C=CN=C1
Molecular Weight (g/mol)178.21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Bright yellow
Appearance (Form)Powder
Titration with HClO4>=88.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only