Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 168552500
Price (USD)/ Each
555.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
654.00 
Save 98.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
555.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
654.00 
Save 98.35 (15%)
Add to cart
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
Catalog number168552500
Price (USD)/ Each
555.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
654.00 
Save 98.35 (15%)
-
Add to cart
Chemical Identifiers
CAS25895-60-7
IUPAC Namesodium cyanoboranuide
Molecular FormulaCH3BNNa
InChI KeyCVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES[Na+].[BH3-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only