Thermo Scientific Chemicals

Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 168575000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Hexanes, 95+%, for spectroscopy, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 168575000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 168575000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=95.0 %
GC
(95% n-hexane approximately)
Water
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text