Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
25 mL
1 L
2.5 L
25 L
Catalog number 168600100
Price (USD)/ Each
269.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
317.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
10 L
Request bulk or custom format
Price (USD)/ Each
269.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
317.00 
Save 47.35 (15%)
Add to cart
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Catalog number168600100
Price (USD)/ Each
269.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
317.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Solution
Strength5.3 to 5.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only