D(+)-Tryptophan, 99%
D(+)-Tryptophan, 99%
Thermo Scientific Chemicals

D(+)-Tryptophan, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 168630050
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.60 
Save 6.95 (14%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.60 
Save 6.95 (14%)
Add to cart
D(+)-Tryptophan, 99%
Catalog number168630050
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.60 
Save 6.95 (14%)
-
Add to cart
Chemical Identifiers
CAS153-94-6
IUPAC Name(2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular FormulaC11H12N2O2
InChI KeyQIVBCDIJIAJPQS-SECBINFHSA-N
SMILESN[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only