Thermo Scientific Chemicals

D(+)-Tryptophan, 99%

Catalog number: 168631000
100 g, Each
Thermo Scientific Chemicals

D(+)-Tryptophan, 99%

Catalog number: 168631000
100 g, Each
Quantity
Catalog number: 168631000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
153-94-6
IUPAC Name
(2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Formula
C11H12N2O2
InChI Key
QIVBCDIJIAJPQS-SECBINFHSA-N
SMILES
N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %
Loss on drying
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text