Thermo Scientific Chemicals

6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 168690250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 168690250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 168690250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5326-23-8
IUPAC Name
6-chloropyridine-3-carboxylic acid
Molecular Formula
C6H4ClNO2
InChI Key
UAWMVMPAYRWUFX-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Cl)N=C1
Appearance (Color)
Off-white to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
186°C to 190°C
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text