Thermo Scientific Chemicals

Hydantoin, 99%, Thermo Scientific Chemicals

Catalog number: 168701000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydantoin, 99%, Thermo Scientific Chemicals

Catalog number: 168701000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 168701000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
461-72-3
IUPAC Name
imidazolidine-2,4-dione
Molecular Formula
C3H4N2O2
InChI Key
WJRBRSLFGCUECM-UHFFFAOYSA-N
SMILES
O=C1CNC(=O)N1
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
220°C to 223°C
Titration with NaOH
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text