2-(Ethylamino)ethanol, 98%, Thermo Scientific Chemicals
2-(Ethylamino)ethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Ethylamino)ethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 168732500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-73-6
IUPAC Name2-(ethylamino)ethan-1-ol
Molecular FormulaC4H11NO
InChI KeyMIJDSYMOBYNHOT-UHFFFAOYSA-N
SMILESCCNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only