4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
4-Dimethylaminobenzaldehyde, 99+%
Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, 99+%

CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 168760250
Price (USD)
24.65
Special offer
Online exclusive
Ends: 26-Sep-2025
30.50
Save 5.85 (19%)
Each
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 26-Sep-2025
30.50
Save 5.85 (19%)
Each
Add to cart
Chemical Identifiers
CAS100-10-7
IUPAC Name4-(dimethylamino)benzaldehyde
Molecular FormulaC9H11NO
InChI KeyBGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point72°C to 75°C
Appearance (Color)Light yellow to light green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link