1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 168831000
Price (USD)/ Each
45.65
Online exclusive
50.70 
Save 5.05 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.70 
Save 5.05 (10%)
Add to cart
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
Catalog number168831000
Price (USD)/ Each
45.65
Online exclusive
50.70 
Save 5.05 (10%)
-
Add to cart
Chemical Identifiers
CAS78-77-3
IUPAC Name1-bromo-2-methylpropane
Molecular FormulaC4H9Br
InChI KeyHLVFKOKELQSXIQ-UHFFFAOYSA-N
SMILESCC(C)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4340 to 1.4370 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only