2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 168871000
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC8H19NO5
InChI KeyOWMVSZAMULFTJU-UHFFFAOYSA-N
SMILESOCCN(CCO)C(CO)(CO)CO
Molecular Weight (g/mol)209.24
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder
Water=<1.0 % (K.F.)
Titration with HClO4>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only