Thermo Scientific Chemicals

Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals

Catalog number: 168885000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals

Catalog number: 168885000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 168885000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-47-4
IUPAC Name
potassium 2-methylpropan-2-olate
Molecular Formula
C4H9KO
InChI Key
LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES
[K+].CC(C)(C)[O-]
KOH + K2CO3 (as KOH)
=<0.50 %
Appearance (Form)
Powder
Appearance (Color)
White to off-white
Titration with HCl
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text