Thermo Scientific Chemicals

(+)-Diethyl L-tartrate, 99+%, Thermo Scientific Chemicals

Catalog number: 168965000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

(+)-Diethyl L-tartrate, 99+%, Thermo Scientific Chemicals

Catalog number: 168965000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 168965000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-91-2
IUPAC Name
1,4-diethyl 2,3-dihydroxybutanedioate
Molecular Formula
C8H14O6
InChI Key
YSAVZVORKRDODB-UHFFFAOYNA-N
SMILES
CCOC(=O)C(O)C(O)C(=O)OCC
Color scale
=<50 APHA
GC
>=99.0 %
Appearance (Form)
Clear viscous liquid
Refractive index
1.4450 to 1.4470 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text