Thermo Scientific Chemicals

N-Carbobenzyloxy-L-alanine, 98%

Catalog number: 169000250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Carbobenzyloxy-L-alanine, 98%

Catalog number: 169000250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169000250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1142-20-7
IUPAC Name
(2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Formula
C11H13NO4
InChI Key
TYRGLVWXHJRKMT-QMMMGPOBSA-N
SMILES
C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Specific optical rotation
-14° to -16° (24°C, 589 nm) (c=2, AcOH)
HPLC
>=97.5 %
Appearance of solution
(5% soln. in MeOH) clear, colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text