2,6-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals
2,6-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 169010100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS632-46-2
IUPAC Name2,6-dimethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyHCBHQDKBSKYGCK-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Melting point114°C to 117°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only