Thermo Scientific Chemicals

Lauroyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169030500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Lauroyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169030500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169030500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-16-3
IUPAC Name
dodecanoyl chloride
Molecular Formula
C12H23ClO
InChI Key
NQGIJDNPUZEBRU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(Cl)=O
Appearance (Form)
Clear liquid
GC
>=97.5 %
Color scale
=<60 APHA
Refractive index
1.4440 to 1.4460 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text