Hexanoyl chloride, 97%
Hexanoyl chloride, 97%
Hexanoyl chloride, 97%
Hexanoyl chloride, 97%
Thermo Scientific Chemicals

Hexanoyl chloride, 97%

CAS: 142-61-0 | C6H11ClO | 134.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 169045000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-61-0
IUPAC Namehexanoyl chloride
Molecular FormulaC6H11ClO
InChI KeyYWGHUJQYGPDNKT-UHFFFAOYSA-N
SMILESCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
GC>=96.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link