Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 169050250
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.70 
Save 4.05 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.70 
Save 4.05 (15%)
Add to cart
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Catalog number169050250
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.70 
Save 4.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecyclohexanecarbonyl chloride
Molecular FormulaC7H11ClO
InChI KeyRVOJTCZRIKWHDX-UHFFFAOYSA-N
SMILESClC(=O)C1CCCCC1
Molecular Weight (g/mol)146.61
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to straw yellow
Infrared spectrumConforms
Refractive index1.4680 to 1.4700 (20°C, 589 nm)
Titration RCOOH=<2 %
Free acid (HCl)Traces
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only