Thermo Scientific Chemicals

Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169051000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169051000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169051000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2719-27-9
IUPAC Name
cyclohexanecarbonyl chloride
Molecular Formula
C7H11ClO
InChI Key
RVOJTCZRIKWHDX-UHFFFAOYSA-N
SMILES
ClC(=O)C1CCCCC1
Titration RCOCl
>=97.5 %
GC
>=97.5 %
Appearance (Color)
Clear colorless to straw yellow
Infrared spectrum
Conforms
Refractive index
1.4680 to 1.4700 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text