Thermo Scientific Chemicals

L(+)-Homoarginine hydrochloride, 98%

Catalog number: 169090050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

L(+)-Homoarginine hydrochloride, 98%

Catalog number: 169090050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169090050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1483-01-8
IUPAC Name
hydrogen 2-amino-6-[(diaminomethylidene)amino]hexanoic acid chloride
Molecular Formula
C7H17ClN4O2
InChI Key
YMKBVNVCKUYUDM-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC(CCCCN=C(N)N)C(O)=O
Specific optical rotation
+17° to +19° (20°C, 589 nm) (c=1, 1 N HCl)
Appearance (Form)
Granular powder
Titration with HClO4
>=97.5 % (On dried substance)
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text