Thermo Scientific Chemicals

Valeryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169121000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Valeryl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169121000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169121000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
638-29-9
IUPAC Name
pentanoyl chloride
Molecular Formula
C5H9ClO
InChI Key
XGISHOFUAFNYQF-UHFFFAOYSA-N
SMILES
CCCCC(Cl)=O
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4190 to 1.4230 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text