Diethyl trans-1,2-cyclopropanedicarboxylate, 97%, Thermo Scientific Chemicals
Diethyl trans-1,2-cyclopropanedicarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl trans-1,2-cyclopropanedicarboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 169180050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,2-diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Molecular FormulaC9H14O4
InChI KeySXLDHZFJMXLFJU-RNFRBKRXSA-N
SMILESCCOC(=O)[C@@H]1C[C@H]1C(=O)OCC
Molecular Weight (g/mol)186.21
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4390 to 1.4420 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only