Phenanthridine, 98%, Thermo Scientific Chemicals
Phenanthridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenanthridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169250050
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
Add to cart
Phenanthridine, 98%, Thermo Scientific Chemicals
Catalog number169250050
Price (USD)/ Each
115.65
Online exclusive
128.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namephenanthridine
Molecular FormulaC13H9N
InChI KeyRDOWQLZANAYVLL-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C=NC1=CC=CC=C21
Molecular Weight (g/mol)179.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
GC>=97.5 %
Infrared spectrumConforms
Melting point104°C to 108°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only