Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 169260050
Price (USD)/ Each
49.30
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.30
Add to cart
Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals
Catalog number169260050
Price (USD)/ Each
49.30
-
Add to cart
Chemical Identifiers
IUPAC Name2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular FormulaC8F14O3
InChI KeyUFFSXJKVKBQEHC-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol)410.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration by coupling97.5 to 102.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only