Thermo Scientific Chemicals

Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: 169261000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Heptafluorobutyric anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: 169261000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169261000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
336-59-4
IUPAC Name
2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Formula
C8F14O3
InChI Key
UFFSXJKVKBQEHC-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Appearance (Color)
Clear colorless to yellow
Titration by coupling
97.5 to 102.5 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text