Appearance (Color)Colorless to yellow to orange to red to brown
Infrared spectrumConforms
Refractive index1.5290 to 1.5370 (20°C, 589 nm)
GC>=90.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.