Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 169430010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 169430010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 169430010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7705-08-0
IUPAC Name
iron(3+) trichloride
Molecular Formula
Cl3Fe
InChI Key
RBTARNINKXHZNM-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Fe+3]
Appearance (Form)
Powder
Manganese (Mn)
=<3000 ppm
Arsenic (As)
=<20 ppm
Zinc (Zn)
=<1000 ppm
Appearance (Color)
Dark grey to black

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text