Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous

CAS: 7705-08-0 | Cl3Fe | 162.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
1 kg
10 kg
25 kg
Packaging:
View all
Glass Bottle
Catalog number 169430050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7705-08-0
IUPAC Nameiron(3+) trichloride
Molecular FormulaCl3Fe
InChI KeyRBTARNINKXHZNM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Manganese (Mn)=<3000 ppm
Appearance (Color)Dark grey to black
Arsenic (As)=<20 ppm
Zinc (Zn)=<1000 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link