Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus pentachloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169460025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10026-13-8
IUPAC Namepentachloro-λ⁵-phosphane
Molecular FormulaCl5P
InChI KeyUHZYTMXLRWXGPK-UHFFFAOYSA-N
SMILESClP(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or scales
Appearance (Color)Yellow
Titration Argentometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Share catalog number, name or link