Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus tribromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 169471000
Price (USD)/ Each
68.65
Online exclusive
76.00 
Save 7.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
76.00 
Save 7.35 (10%)
Add to cart
Phosphorus tribromide, 99%, Thermo Scientific Chemicals
Catalog number169471000
Price (USD)/ Each
68.65
Online exclusive
76.00 
Save 7.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-60-8
IUPAC Nametribromophosphane
Molecular FormulaBr3P
InChI KeyIPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILESBrP(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)light brown
Appearance (Color)Clear to slightly hazy colorless to yellow to
Titration Argentometric>=98.5 % (ex Br)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only