Thermo Scientific Chemicals

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: 169501000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: 169501000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169501000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2466-76-4
IUPAC Name
1-(1H-imidazol-1-yl)ethan-1-one
Molecular Formula
C5H6N2O
InChI Key
VIHYIVKEECZGOU-UHFFFAOYSA-N
SMILES
CC(=O)N1C=CN=C1
Infrared spectrum
Conforms
Appearance (Form)
Adhering crystalline powder or crystals
Appearance (Color)
White to beige to tan
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text