1,3-Dithiane, 97%, Thermo Scientific Chemicals
1,3-Dithiane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dithiane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169511000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS505-23-7
IUPAC Name1,3-dithiane
Molecular FormulaC4H8S2
InChI KeyWQADWIOXOXRPLN-UHFFFAOYSA-N
SMILESC1CSCSC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point49°C to 54°C
GC>=96.0 %
Appearance (Color)White to light beige
Appearance (Form)Powder or crystals or needles
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only