Thermo Scientific Chemicals

Methyl bromoacetate, 99%, Thermo Scientific Chemicals

Catalog number: 169551000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl bromoacetate, 99%, Thermo Scientific Chemicals

Catalog number: 169551000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169551000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-32-2
IUPAC Name
methyl 2-bromoacetate
Molecular Formula
C3H5BrO2
InChI Key
YDCHPLOFQATIDS-UHFFFAOYSA-N
SMILES
COC(=O)CBr
GC
>=98.5 %
Infrared spectrum
Conforms
Refractive index
1.4570 to 1.4590 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text