Nunc™ Cell Culture/Petri Dishes
Choose the new Thermo Scientific Nunc EasYDish cell culture dishes for improved handling and stacking.
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Cell Culture/Petri Dishes
Thermo Scientific™

Nunc™ Cell Culture/Petri Dishes

Experience optimal performance across a broad range of sizes and styles. Nunc Cell Culture Dishes are designed to suit a variety of experimental needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaSurface TreatmentGriddedVent
16955821.5 sq. cmNunclon DeltaGriddedLid with airvent
1503188.8 sq. cmNunclon DeltaNot GriddedLid without airvent
1530668.8 sq. cmNunclon DeltaNot GriddedLid with airvent
1749268.8 sq. cmNunclon DeltaGriddedLid with airvent
1710998.8 sq. cmNon-treatedNot GriddedLid with airvent
17488820.8 sq. cmCell CultureNot GriddedLid with airvent
15032621.5 sq. cmNunclon DeltaNot GriddedLid without airvent
15028821.5 sq. cmNunclon DeltaNot GriddedLid with airvent
15035056.7 sq. cmNunclon DeltaNot GriddedLid with airvent
168381145 sq. cmNunclon DeltaNot GriddedLid with airvent
17293156.7 sq. cmNunclon DeltaNot GriddedLid with airvent
Catalog number 169558
Price (USD)
601.65
Online Exclusive
644.00
Save 42.35 (7%)
Case of 400
-
Add to cart
Culture Area:
21.5 sq. cm
Surface Treatment:
Nunclon Delta
Gridded:
Gridded
Vent:
Lid with airvent
Price (USD)
601.65
Online Exclusive
644.00
Save 42.35 (7%)
Case of 400
Add to cart

Experience optimal performance across a broad range of sizes and styles to suit a variety of experimental needs. Thermo Scientific Nunc Cell Culture Dishes receive the Nunclon Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types. The Petri dishes are made from high quality and optically clear polystyrene.

Nunc culture dishes, available in a variety of sizes and designs, are made from high quality optically clear polystyrene or Permanox plastic.

  • Top and bottom stacking rings for easier handling and more stable stacking
  • Nunclon Delta-treated dishes are optimized for cell attachment
  • Sterilized by gamma irradiation
  • Available with or without air vents for gas exchange
  • The dishes with 2 x 2 mm grid are ideal for cloning experiments and determination of plating efficiency

Ordering information

Cat. Nos. 174926 and 169558 feature 2mm grids

Cat. Nos. 171099 and 150340 are for suspension culture and not Nunclon-treated

Includes a certificate that assures user of biological performance as well as traceability.

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorClear
Closure IncludedClosure Included
Closure MaterialPlastic, Polystyrene
GriddedGridded
IncludesAirvent
No. per Case400/Cs.
No. per Pack10 Pk.
ShapeRound
TypeCell Culture Dish
VentLid with airvent
Volume (Metric) Working5 mL
Bottom ShapeFlat
Capacity (Metric)5 mL (Well)
ColorClear
Culture Area21.5 sq. cm
Diameter (Metric)60 mm
Height (Metric)15 mm
MaterialPolystyrene
SterilitySterile
Surface TreatmentNunclon Delta
Well Count1
Unit SizeCase of 400

Frequently asked questions (FAQs)

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Share catalog number, name or link