Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals
Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
25 g
1 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 169590020
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals
Catalog number169590020
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7775-14-6
IUPAC Namedisodium sulfinatosulfinate
Molecular FormulaNa2O4S2
InChI KeyJVBXVOWTABLYPX-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=85 %
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only