Thermo Scientific Chemicals

O-Methyl-L-tyrosine, 98%

Catalog number: 169750010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

O-Methyl-L-tyrosine, 98%

Catalog number: 169750010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 169750010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6230-11-1
IUPAC Name
(2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
Molecular Formula
C10H13NO3
InChI Key
GEYBMYRBIABFTA-IMWMWJONNA-N
SMILES
COC1=CC=C(C[C@H](N)C(O)=O)C=C1
Appearance (Color)
White to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %
Specific optical rotation
-7° to -9° (20°C, 589 nm) (c=2, 1 N HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text