Thermo Scientific Chemicals

4-Methylquinoline, 99%, Thermo Scientific Chemicals

Catalog number: 169770250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methylquinoline, 99%, Thermo Scientific Chemicals

Catalog number: 169770250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169770250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
491-35-0
IUPAC Name
4-methylquinoline
Molecular Formula
C10H9N
InChI Key
MUDSDYNRBDKLGK-UHFFFAOYSA-N
SMILES
CC1=C2C=CC=CC2=NC=C1
Appearance (Color)
Clear light yellow to green to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.6180 to 1.6220 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text