Thermo Scientific Chemicals

5-Bromovaleric acid, 97%, Thermo Scientific Chemicals

Catalog number: 169820100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromovaleric acid, 97%, Thermo Scientific Chemicals

Catalog number: 169820100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169820100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2067-33-6
IUPAC Name
5-bromopentanoic acid
Molecular Formula
C5H9BrO2
InChI Key
WNXNUPJZWYOKMW-UHFFFAOYSA-N
SMILES
OC(=O)CCCCBr
Appearance (Color)
Yellow to light brown
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
37°C to 41°C
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text