Thermo Scientific Chemicals

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

Catalog number: 169831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

Catalog number: 169831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169831000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-11-6
IUPAC Name
diiodomethane
Molecular Formula
CH2I2
InChI Key
NZZFYRREKKOMAT-UHFFFAOYSA-N
SMILES
ICI
Appearance (Color)
Clear colorless to yellow to red to orange-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text