Thermo Scientific Chemicals

1,8-Bis(dimethylamino)naphthalene, 99%, Thermo Scientific Chemicals

Catalog number: 169860500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1,8-Bis(dimethylamino)naphthalene, 99%, Thermo Scientific Chemicals

Catalog number: 169860500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169860500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20734-58-1
IUPAC Name
N1,N1,N8,N8-tetramethylnaphthalene-1,8-diamine
Molecular Formula
C14H18N2
InChI Key
GJFNRSDCSTVPCJ-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC2=CC=CC(N(C)C)=C12
Melting point
46°C to 50°C
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
Light yellow to beige to pink or brown
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text