Thermo Scientific Chemicals

Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals

Catalog number: 169880100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals

Catalog number: 169880100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 169880100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
631-61-8
IUPAC Name
ammonium acetate
Molecular Formula
C2H7NO2
InChI Key
USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES
[NH4+].CC([O-])=O
Appearance (Form)
Adhering crystals
Titration with HClO4
>=97.5 % (On anhydrous substance)
Water
=<2 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text