Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 169890100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 169890100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 169890100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1493-13-6
IUPAC Name
trifluoromethanesulfonic acid
Molecular Formula
CHF3O3S
InChI Key
ITMCEJHCFYSIIV-UHFFFAOYSA-N
SMILES
OS(=O)(=O)C(F)(F)F
Titration with NaOH
98.5 to 101.5 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.3260 to 1.3300 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text