Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 mL
10 mL
50 mL
1 L
Packaging:
Glass bottle
Catalog number 169892500
Price (USD)/ Each
597.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
703.00 
Save 105.35 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
597.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
703.00 
Save 105.35 (15%)
Add to cart
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Catalog number169892500
Price (USD)/ Each
597.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
703.00 
Save 105.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1493-13-6
IUPAC Nametrifluoromethanesulfonic acid
Molecular FormulaCHF3O3S
InChI KeyITMCEJHCFYSIIV-UHFFFAOYSA-N
SMILESOS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH98.5 to 101.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3260 to 1.3300 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only