Nalidixic acid, 99.5%, Thermo Scientific Chemicals
Nalidixic acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalidixic acid, 99.5%, Thermo Scientific Chemicals

Nalixidic acid, or nalixidin, CAS # 389-08-2, is a quinolone antibiotic used to inhibit DNA gyrase and prevent cell division.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 169901000
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
Add to cart
Nalidixic acid, 99.5%, Thermo Scientific Chemicals
Catalog number169901000
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
CAS389-08-2
IUPAC Name1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Molecular FormulaC12H12N2O3
InChI KeyMHWLWQUZZRMNGJ-UHFFFAOYSA-N
SMILESCCN1C=C(C(O)=O)C(=O)C2=CC=C(C)N=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with NaOH>=99.4 %
Infrared spectrumConforms
Appearance (Form)Powder
Melting point225°C to 231°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalixidic acid is a synthetic quinolone that is a monocarboxylic acid composed of 1,8-naphthyridine substituted with carboxylic acid, ethyl, and methyl groups

• Nalixidic acid is a DNA gyrase inhibitor that prevents DNA synthesis and subsequent cell divisions

Applications

• Nalixidic acid is primarily effective against gram-negative bacteria

• It may be useful for bacterial resistance studies and laboratory research on drug allergies and cross-reactivity

• Nalixidic acid is suitable for high-performance liquid chromatography (HPLC) and related biochemistry laboratory applications

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Kumar, Y.; Sharma, A.; Mani, K.R. Characterization of antimicrobial resistance markers & their stability in Salmonella enterica serovar Typhi. Indian J Med Res. 2017, 146, S9-S14.
  2. Morales-Irigoyen, E.E.; de Las Mercedes Gómez-Y-Gómez, Y.; Flores-Moreno, J.L.; Franco-Hernández, M.O. A bionanohybrid ZnAl-NADS ecological pesticide as a treatment for soft rot disease in potato (Solanum tuberosum L.). Environ Sci Pollut Res Int. 2018, 25(22), 21430-21439.