Thermo Scientific Chemicals

Phenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169940500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 169940500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169940500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
701-99-5
IUPAC Name
2-phenoxyacetyl chloride
Molecular Formula
C8H7ClO2
InChI Key
PKUPAJQAJXVUEK-UHFFFAOYSA-N
SMILES
ClC(=O)COC1=CC=CC=C1
Appearance (Color)
Clear light yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration RCOCl
>=97.5 %
Refractive index
1.5324 to 1.5344 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text