Thermo Scientific Chemicals

1-Triphenylphosphoranylidene-2-propanone, 99%, Thermo Scientific Chemicals

Catalog number: 169971000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Triphenylphosphoranylidene-2-propanone, 99%, Thermo Scientific Chemicals

Catalog number: 169971000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1439-36-7
IUPAC Name
1-(triphenyl-λ⁵-phosphanylidene)propan-2-one
Molecular Formula
C21H19OP
InChI Key
KAANTNXREIRLCT-UHFFFAOYSA-N
SMILES
CC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
203°C to 211°C
HPLC
>=98.5 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text