Thermo Scientific Chemicals

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 169980500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 169980500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 169980500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1067-74-9
IUPAC Name
methyl 2-(diethoxyphosphoryl)acetate
Molecular Formula
C7H15O5P
InChI Key
CTSAXXHOGZNKJR-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC(=O)OC)OCC
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4310 to 1.4350 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text