N-(2,3-Epoxypropyl)phthalimide, 95%
N-(2,3-Epoxypropyl)phthalimide, 95%
N-(2,3-Epoxypropyl)phthalimide, 95%
N-(2,3-Epoxypropyl)phthalimide, 95%
Thermo Scientific Chemicals

N-(2,3-Epoxypropyl)phthalimide, 95%

CAS: 5455-98-1 | C11H9NO3 | 203.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
View all
Glass Bottle
Catalog number 170040100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS5455-98-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point92°C to 102°C
GC>=94.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
345034Certificate of AnalysisFeb 08, 2023170040250, 170040100
344531Certificate of AnalysisJan 23, 2023170040250, 170040100
343857Certificate of AnalysisDec 15, 2022170040250, 170040100
344724Certificate of AnalysisDec 10, 2021170040250, 170040100
344533Certificate of AnalysisApr 28, 2021170040250, 170040100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection